Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SGK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114778
Description
Antibody detects endogenous levels of total SGK3.
SGKL (SGK3) is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK). Unlike SGK, SGKL is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| SGK3 | |
| Polyclonal | |
| Unconjugated | |
| SGK3 | |
| 2510015P22Rik; A330005P07Rik; CISK; cytokine-independent survival kinase; DKFZp781N0293; fy; fz; Serine/threonine-protein kinase Sgk3; serum glucocorticoid regulated kinase 3; serum/glucocorticoid regulated kinase 3; serum/glucocorticoid regulated kinase family member 3; serum/glucocorticoid regulated kinase family, member 3; serum/glucocorticoid-regulated kinase 3; Serum/glucocorticoid-regulated kinase-like; SGK2; SGK3; Sgkl | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170755, 171498, 23678 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8R4V0, Q96BR1, Q9ERE3 | |
| SGK3 | |
| A synthesized peptide derived from human SGK3(Accession Q96BR1), corresponding to amino acid residues Y460-L496. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction