Learn More
Invitrogen™ SGK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533021
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Bat, Horse, Rabbit (95%); Elephant, Panda, Opossum, Turkey, Chicken (90%); Platypus, Lizard, Pufferfish, Zebrafish (85%); Xenopus (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
SGKL (SGK3) is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK). Unlike SGK, SGKL is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
SGK3 | |
Polyclonal | |
Unconjugated | |
SGK3 | |
2510015P22Rik; A330005P07Rik; CISK; cytokine-independent survival kinase; DKFZp781N0293; fy; fz; Serine/threonine-protein kinase Sgk3; serum glucocorticoid regulated kinase 3; serum/glucocorticoid regulated kinase 3; serum/glucocorticoid regulated kinase family member 3; serum/glucocorticoid regulated kinase family, member 3; serum/glucocorticoid-regulated kinase 3; Serum/glucocorticoid-regulated kinase-like; SGK2; SGK3; Sgkl | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23678 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q96BR1 | |
SGK3 | |
Synthetic 20 amino acid peptide from N-terminus of human SGK3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.