Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGK494 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983956
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3956 Supplier Bioss Supplier No. BS16116R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLJ25006 likely exerts a protein serine/threonine kinase activity.

Specifications

Antigen SGK494
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SGK494
Gene Accession No. Q96LW2
Gene Alias BC030499; BC037104; cDNA sequence BC030499; Sgk494; sugen kinase 494; uncharacterized serine/threonine-protein kinase SgK494
Gene Symbols SGK494
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FLJ25006.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124923
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.