Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGLT2 Rabbit anti-Human, Mouse, Non-human Primate, Polyclonal, FabGennix

Catalog No. 502209097
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9097 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9097 Supplier Fabgennix Inc Supplier No. SGLT2201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SGLT2 Polyclonal antibody specifically detects SGLT2 in Human, Mouse, Non-human Primate samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen SGLT2
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene SLC5A2
Gene Accession No. P31639, Q923I7
Gene Alias glucose transporter; bSGLT2; low affinity Na-dependent glucose transporter (SGLT2); low affinity sodium-dependent glucose cotransporter; low affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 2; Na(+)/nucleoside cotransporter; Na+/glucose cotransporter 2; Na+/nucleoside cotransporter; Sglt2; SGLT-2; SLC5A2; SNST1; sodium/glucose cotransporter 2; Sodium/nucleoside cotransporter; sodium-glucose cotransporter-like 2; solute carrier family 5 (sodium/glucose cotransporter), member 2; solute carrier family 5 (sodium/glucose transporter), member 2; solute carrier family 5 member 2
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 580-630 on human SGLT2 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246787, 467963, 6524
Target Species Human, Mouse, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.