Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGLT2/SLC5A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$159.60 - $391.00
Specifications
Antigen | SGLT2/SLC5A2 |
---|---|
Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SGLT2/SLC5A2 Polyclonal antibody specifically detects SGLT2/SLC5A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
SGLT2/SLC5A2 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Tumor Suppressors | |
PBS, 50% glycerol, pH7.3 | |
6524 | |
IgG | |
Affinity purified |
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Low affinity sodium-glucose cotransporter, Na(+)/glucose cotransporter 2, SGLT2sodium/glucose cotransporter 2, solute carrier family 5 (sodium/glucose cotransporter), member 2, solute carrier family 5 (sodium/glucose transporter), member 2, Solute carrier family 5 member 2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 564-624 of human SGLT2/SLC5A2 (NP_003032.1). RHSKEEREDLDADEQQGSSLPVQNGCPESAMEMNEPQAPAPSLFRQCLLWFCGMSRGGVGS | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title