Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGLT3/SLC5A4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159886
Description
SGLT3/SLC5A4 Polyclonal specifically detects SGLT3/SLC5A4 in Human samples. It is validated for Western Blot.Specifications
SGLT3/SLC5A4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DJ90G24.4, low affinity sodium-glucose cotransporter, Na(+)/glucose cotransporter 3, SAAT1low affinity sodium glucose cotransporter, SGLT2, SGLT3, sodium transporter, Sodium/glucose cotransporter 3, solute carrier family 5 (low affinity glucose cotransporter), member 4, solute carrier family 5 (neutral amino acid transporters, system A), member 4, Solute carrier family 5 member 4 | |
Rabbit | |
72 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Dusky titi monkey: 84%;. | |
Human | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9NY91 | |
SLC5A4 | |
Synthetic peptides corresponding to SLC5A4(solute carrier family 5 (low affinity glucose cotransporter), member 4) (NP_055042). The peptide sequence was selected from the N terminal of SLC5A4. Peptide sequence MASTVSPSTIAETPEPPPLSDHIRNAADISVIVIYFLVVMAVGLWAMLKT The peptide sequence for this immunogen was taken from within the described region. | |
Protein A purified | |
RUO | |
6527 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction