Learn More
Invitrogen™ SGMS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590177
Description
Immunogen sequence: MKEVVYWSPK KVADWLLENA MPEYCEPLEH FTGQDLINLT QEDFKKPPLC RVSSDNGQRL LDMIETLKME HHLEAHKNGH ANGHLNIGVD IPTPDGSFSI KIKPNGMPNG YRKEMIKIPM PELERSQYPM EWGK; Positive Samples: A-549; Cellular Location: Golgi apparatus membrane, Multi-pass membrane protein.
A bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and the inverse reaction. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Directly and specifically recognizes the choline head group on the substrate. Also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. May not function strictly as a SM synthase. Inhibited by bacterial PC-phospholipase C inhibitor D609. Localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. Possibly present on Brain, heart, kidney, liver, muscle and stomach.
Specifications
SGMS1 | |
Polyclonal | |
Unconjugated | |
SGMS1 | |
9530058O11Rik; AI841905; C80702; hmob33; Medulla oblongata-derived protein; MGC17342; MOB; MOB1; mob-1; Phosphatidylcholine:ceramide cholinephosphotransferase 1; protein Mob; SGMS1; SMS1; Sor1; sphingomyelin synthase 1; TMEM23; Transmembrane protein 23 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
259230, 353229 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.9 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q7TSX5, Q86VZ5 | |
SGMS1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-134 of human SGMS1 (NP_6715121). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.