Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGOL1, Mouse anti-Human, Clone: 3C11, Abnova™

Catalog No. 89968810 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-810 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-968-810 Supplier Abnova Corporation Supplier No. H00151648M01
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sequence: MAKERCLKKSFQDSLEDIKKRMKEKRNKNLAEIGKRRSFIAAPCQIITNTSTLLKNYQDNNKMLVLALENEKSKVKEAQDIILQLRKECYYLTCQLYALKGKLTSQQTVEPAQNQEICSSGMDPNSDDSSRNLFVKDLPQIPLEETELPGQGESFQIEATPPETQQSPHLSLKDITNVSLYPVVKIRRLSLSPKKNKASPAVALPKRRCTASVNYKEPTLASKLRRGDPFTDLCFLNSPIFKQKKDLRRSKKRALEVSPAKEAIFILYYVREFVSRFPDCRKCKLETHICLR

Specifications

Antigen SGOL1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3C11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene shugoshin-like 1 (S. pombe)
Gene Accession No. BC017867
Gene Alias NY-BR-85/SGO/Sgo1
Gene Symbols SGOL1
Host Species Mouse
Immunogen SGOL1 (AAH17867, 1 a.a. ∼ 292 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151648
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.