Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGOL2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060454DL594
Description
SGOL2 Polyclonal antibody specifically detects SGOL2 in Human samples. It is validated for Western BlotSpecifications
SGOL2 | |
Polyclonal | |
Western Blot | |
FLJ25211, MGC129665, Sgo2, Shugoshin-2, shugoshin-like 2 (S. pombe), Tripin, TRIPINshugoshin-like 2 | |
The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human shugoshin-like 2 using the numbering given in entry NP_689737.3 (GeneID 151246). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
151246 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction