Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SGPP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543467
Description
Peptide sequence: THKYAPFIII GLHLALGIFS FTLDTWSTSR GDTAEILGSG AGIACGSHVT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Sphingosine 1-phosphate (S1P) is a highly bioactive lipid that has a myriad of biological effects both intracellularly as a second messenger and extracellularly by binding to the S1P(1-5)/G-protein-coupled receptors of the endothelial differentiation gene family. Intracellularly, at least two enzymes, sphingosine kinase (1 and 2) and S1P phosphatase, regulate the activity of S1P by governing the phosphorylation of S1P. Regulation of hS1Pase1 with the resultant changes in cellular and secreted S1P has important implications in cell proliferation, angiogenesis and apoptosis.
Specifications
SGPP1 | |
Polyclonal | |
Unconjugated | |
SGPP1 | |
AI463453; hSPP1; hSPPase1; Sgpp1; Sphingosine-1-phosphatase 1; sphingosine-1-phosphate phosphatase 1; sphingosine-1-phosphate phosphohydrolase; Sphingosine-1-phosphate phosphohydrolase 1; SPP; SPP1; SPP-1; SPPase1; Spph1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
81537 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BX95 | |
SGPP1 | |
Synthetic peptide directed towards the middle region of human SGPP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction