Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGPP1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA562741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562741 Supplier Thermo Scientific Supplier No. PA562741

Rabbit Polyclonal Antibody

Immunogen sequence: KITIPLACKI FNIPCDDIRK ARQHMEVELP YR Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.

Sphingosine 1-phosphate (S1P) is a highly bioactive lipid that has a myriad of biological effects both intracellularly as a second messenger and extracellularly by binding to the S1P(1-5)/G-protein-coupled receptors of the endothelial differentiation gene family. Intracellularly, at least two enzymes, sphingosine kinase (1 and 2) and S1P phosphatase, regulate the activity of S1P by governing the phosphorylation of S1P. Regulation of hS1Pase1 with the resultant changes in cellular and secreted S1P has important implications in cell proliferation, angiogenesis and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGPP1
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SGPP1
Gene Accession No. Q9BX95
Gene Alias AI463453; hSPP1; hSPPase1; Sgpp1; Sphingosine-1-phosphatase 1; sphingosine-1-phosphate phosphatase 1; sphingosine-1-phosphate phosphohydrolase; Sphingosine-1-phosphate phosphohydrolase 1; SPP; SPP1; SPP-1; SPPase1; Spph1
Gene Symbols SGPP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SGPP1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81537
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.