Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGSH/Sulphamidase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981643
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1643 Supplier Bioss Supplier No. BS11756R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SGSH is one of several enzymes involved in the lysosomal degradation of heparan sulfate. Mutations in this gene are associated with Sanfilippo syndrome A, one type of the lysosomal storage disease mucopolysaccaridosis III, which results from impaired degradation of heparan sulfate. Transcripts of varying sizes have been reported but their biological validity has not been determined.

Specifications

Antigen SGSH/Sulphamidase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SGSH
Gene Accession No. P51688
Gene Alias 4632406A19Rik; heparan sulfate sulfatase; HSS; MPS3A; mucopolysaccharidosis type IIIA; N-sulfoglucosamine sulfohydrolase; N-sulfoglucosamine sulfohydrolase (sulfamidase); N-sulphoglucosamine sulphohydrolase; SFMD; SGSH; Sulfoglucosamine sulfamidase; Sulphamidase
Gene Symbols SGSH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Sulphamidase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6448
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.