Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGSM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534438
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534438 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534438 Supplier Thermo Scientific Supplier No. PA534438
Add to Cart
Add to Cart

SGSM1 Polyclonal Antibody

SGSM1 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen SGSM1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SGSM1
Gene Accession No. Q2NKQ1, Q8BPQ7
Gene Alias 2410098H20Rik; AA408781; D5Bwg1524e; E130107M15; KIAA1941; NuIP; nurr1-interacting protein; RUN and TBC1 domain containing 2; RUN and TBC1 domain-containing protein 2; RUTBC2; SGSM1; Small G protein signaling modulator 1; small G protein signaling modulator 1 protein
Gene Symbols SGSM1
Host Species Rabbit
Immunogen A 17 amino acid synthetic peptide near the center of human SGSM1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129049, 288743, 52850
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.