Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGSM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555106 Supplier Invitrogen™ Supplier No. PA555106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HRDSTISNDV FISVDDLEPP EPQDPEDSRP KPEQEAGPGT PGTAVVEQQH SVEFDSPDSG LPSSRNYSVA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.

Small G proteins such as RAP and RAB proteins are the key molecules in intracellular signal transduction and vesicle transportation. A novel protein family small G protein signaling modulator (SGSM) consisting of three members SGSM1-3 bind to RAP and RAB family proteins. All three SGSM proteins possess both a RUN domain and a TBC domain. SGSM2 is a Rab9A effector that activates GTP hydrolysis by Rab32 and Rab33B proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGSM2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SGSM2
Gene Accession No. O43147
Gene Alias D630003G22Rik; KIAA0397; mKIAA0397; R74628; RUN and TBC1 domain containing 1; RUN and TBC1 domain-containing protein 1; RUTBC1; Sgsm2; Small G protein signaling modulator 2; small G protein signaling modulator 2 protein
Gene Symbols SGSM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SGSM2. Recombinant protein control fragment (Product #RP-93280).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9905
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.