Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGSM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534439
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534439 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534439 Supplier Invitrogen™ Supplier No. PA534439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human cerebellum tissue lysate. PA5-34439 can be used with blocking peptide PEP-1481.

Small G proteins such as RAP and RAB proteins are the key molecules in intracellular signal transduction and vesicle transportation. A novel protein family small G protein signaling modulator (SGSM) consisting of three members SGSM1-3 bind to RAP and RAB family proteins. All three SGSM proteins possess both a RUN domain and a TBC domain. SGSM2 is a Rab9A effector that activates GTP hydrolysis by Rab32 and Rab33B proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGSM2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SGSM2
Gene Accession No. O43147, Q80U12
Gene Alias D630003G22Rik; KIAA0397; mKIAA0397; R74628; RUN and TBC1 domain containing 1; RUN and TBC1 domain-containing protein 1; RUTBC1; Sgsm2; Small G protein signaling modulator 2; small G protein signaling modulator 2 protein
Gene Symbols SGSM2
Host Species Rabbit
Immunogen An 18 amino acid synthetic peptide near the center of human SGSM2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303304, 97761, 9905
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.