Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGSM3 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297659MFV450
Description
SGSM3 Polyclonal antibody specifically detects SGSM3 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
SGSM3 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
DJ1042K10.2, DKFZp761D051, MAP, merlin binding protein, Merlin-associated protein, rabGAPLP, Rab-GTPase-activating protein-like protein, RUN and SH3 containing 3, RUN and TBC1 domain containing 3, RUN and TBC1 domain-containing protein 3, RUSC3, RUTBC3, small G protein signaling modulator 3, small G protein signaling modulator 3 protein | |
Produced in rabbits immunized with E. coli-derived Human SGSM3 fragment. | |
0.1 mL | |
Signal Transduction | |
27352 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction