Learn More
SGSM3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305661AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Small G proteins such as RAP and RAB proteins are the key molecules in intracellular signal transduction and vesicle transportation. A novel protein family small G protein signaling modulator (SGSM) consisting of three members SGSM1-3 bind to RAP and RAB family proteins. All three SGSM proteins possess both a RUN domain and a TBC domain. Little is known of the exact function of SGSM3.Specifications
SGSM3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8VCZ6, Q96HU1 | |
SGSM3 | |
Between 699 and 749 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
SGSM3 | |
SGSM3; small G protein signaling modulator 3; DKFZp761D051; MAP; RUSC3; RUTBC3; RUN and SH3 containing 3; RUN and TBC1 domain containing 3; merlin binding protein; merlin-associated protein; small G protein signaling modulator 3 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
105835, 27352 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.