Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGTA Antibody (19E8), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259426AF750
This item is not returnable.
View return policy
Description
SGTA Monoclonal antibody specifically detects SGTA in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
| SGTA | |
| Monoclonal | |
| Alexa Fluor 750 | |
| alphaSGT, Alpha-SGT, protein containing three tetratricopeptide repeats, SGT1, SGThSGT, small glutamine-rich tetratricopeptide repeat (TPR)-containing, small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha, small glutamine-rich tetratricopeptide repeat-containing protein alpha, UBP, Vpu-binding protein | |
| Recombinant human SGTA (1-313aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| 6449 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 19E8. | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction