Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGTA Antibody (19E8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259426H
This item is not returnable.
View return policy
Description
SGTA Monoclonal antibody specifically detects SGTA in Human samples. It is validated for Western Blot, ELISASpecifications
SGTA | |
Monoclonal | |
HRP | |
alphaSGT, Alpha-SGT, protein containing three tetratricopeptide repeats, SGT1, SGThSGT, small glutamine-rich tetratricopeptide repeat (TPR)-containing, small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha, small glutamine-rich tetratricopeptide repeat-containing protein alpha, UBP, Vpu-binding protein | |
Recombinant human SGTA (1-313aa) purified from E. coli | |
0.1 mL | |
Apoptosis, Cell Biology, Cell Cycle and Replication, Signal Transduction | |
6449 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
19E8. | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction