Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGTA Monoclonal Antibody (6A4)
GREENER_CHOICE

Catalog No. PIMA531545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531545 Supplier Invitrogen™ Supplier No. MA531545
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Positive Control: 293T, A431, HeLa, HepG2, U87-MG, SK-N-SH, IMR32, SK-N-AS, SGT-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGTA
Applications Western Blot
Classification Monoclonal
Clone 6A4
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SGTA
Gene Accession No. O43765
Gene Alias 5330427H01Rik; AI194281; alpha h UBP; alphaSGT; Alpha-SGT; D10Ertd190e; hSGT; protein containing three tetratricopeptide repeats; Sgt; Sgt1; SGTA; small glutamine rich tetratricopeptide repeat containing alpha; small glutamine-rich protein with tetratricopeptide repeats 1; small glutamine-rich tetratricopeptide repeat (TPR) containing protein; small glutamine-rich tetratricopeptide repeat (TPR) containing protein (SGT); small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha; small glutamine-rich tetratricopeptide repeat-containing protein alpha; Stg; UBP; vpu-binding protein
Gene Symbols SGTA
Host Species Mouse
Immunogen The immunogen used to generate this antibody corresponds to human SGTA.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6449
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.