Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SGTB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564128
Description
Immunogen sequence: EQLRNHIRSR SFSSSAEEHS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
SGTB is co-chaperone that binds directly to HSC70 and HSP70 and regulates their ATPase activity.
Specifications
SGTB | |
Polyclonal | |
Unconjugated | |
SGTB | |
Beta-SGT; C630001O05Rik; SGT2; Sgtb; small glutamine rich protein with tetratricopeptide repeats 2; small glutamine rich tetratricopeptide repeat containing beta; small glutamine-rich protein with tetratricopeptide repeats 2; small glutamine-rich tetratricopeptide repeat (TPR)-containing, beta; small glutamine-rich tetratricopeptide repeat-containing protein beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54557 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96EQ0 | |
SGTB | |
Recombinant protein corresponding to Human SGTB. Recombinant protein control fragment (Product #RP-103519). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction