Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGTB Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571185
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1185 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-1185 Supplier Bethyl Laboratories, Inc Supplier No. A305771AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

SGTB is co-chaperone that binds directly to HSC70 and HSP70 and regulates their ATPase activity.

Specifications

Antigen SGTB
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SGTB
Gene Accession No. Q96EQ0
Gene Alias SGT2, beta-SGT, small glutamine rich protein with tetratricopeptide repeats 2, small glutamine-rich protein with tetratricopeptide repeats 2, small glutamine-rich tetratricopeptide repeat-containing protein beta
Gene Symbols SGTB
Host Species Rabbit
Immunogen between 254 and 304
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54557
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.