Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SGX 523
GREENER_CHOICE

Catalog No. 535610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-561-0 10 mg
53-565-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-561-0 Supplier Tocris Bioscience™ Supplier No. 5356/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective and potent c-MET kinase inhibitor

SGX 523 is a potent and selective c-MET kinase inhibitor (IC50 = 4nM). Exhibits minimal inhibition against a panel of 213 other protein kinases (at 1 μM). Suppresses growth of tumor xenografts derived from human glioblastoma, lung and gastric cancer cells.

Specifications

Quantity 10 mg
Product Type SGX 523
Purity >99%
Molecular Weight (g/mol) 359.41

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.