Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH2B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578460 Supplier Invitrogen™ Supplier No. PA578460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: C2C12, PC-12, Rat-2, mouse brain, rat brain Predicted Reactivity: Bovine (96%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the SH2-domain containing mediators family. The encoded protein mediates activation of various kinases and may function in cytokine and growth factor receptor signaling and cellular transformation. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2B1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SH2B1
Gene Accession No. Q62985, Q91ZM2, Q9NRF2
Gene Alias AI425885; C530001K22Rik; DKFZp547G1110; fceRI gamma-chain-interacting protein SH2-B; FLJ30542; Irip; KIAA1299; mKIAA1299; pro-rich, PH and SH2 domain-containing signaling mediator; PSM; SH2 B; SH2 domain-containing protein 1B; SH2 domain-containing putative adapter SH2-B; Sh2b; SH2-B; SH2B adapter protein 1; SH2B adaptor protein 1; SH2-B PH domain containing signaling mediator 1; SH2-B PH domain-containing signaling mediator 1; SH2-B signaling protein; SH2B1; SH2B1 protein; SH2-Bb; Sh2bpsm1
Gene Symbols SH2B1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SH2B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20399, 25970, 89817
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.