Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH2B3 Monoclonal Antibody (OTI2B11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502748
Description
This gene encodes a member of the SH2B adaptor family of proteins, which are involved in a range of signaling activities by growth factor and cytokine receptors. The encoded protein is a key negative regulator of cytokine signaling and plays a critical role in hematopoiesis. Mutations in this gene have been associated with susceptibility to celiac disease type 13 and susceptibility to insulin-dependent diabetes mellitus.Specifications
| SH2B3 | |
| Monoclonal | |
| 0.61 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UQQ2 | |
| SH2B3 | |
| Full length human recombit protein of human SH2B3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2B11 | |
| Unconjugated | |
| SH2B3 | |
| IDDM20, LNK | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10019 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction