Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH2D1A Antibody (028), Alexa Fluor™ 750 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289820AF750
Description
SH2D1A Monoclonal antibody specifically detects SH2D1A in Human samples. It is validated for ELISASpecifications
SH2D1A | |
Monoclonal | |
Alexa Fluor 750 | |
DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLPDFLJ18687, XLPSH2 domain protein 1A | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human SH2D1A (NP_002342.1; Pro2-Lys97). | |
0.1 mL | |
Immune System Diseases, Immunology | |
4068 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
028 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction