Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH2D1A Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299726APC
Description
SH2D1A Polyclonal antibody specifically detects SH2D1A in Human samples. It is validated for ELISASpecifications
SH2D1A | |
Polyclonal | |
ELISA | |
DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLPDFLJ18687, XLPSH2 domain protein 1A | |
Produced in rabbits immunized with purified, recombinant Human SH2D1A (Accession#: NP_002342.1; Pro2-Lys97) | |
0.1 mL | |
Immune System Diseases, Immunology | |
4068 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction