Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SH2D1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145022
Description
Immunogen sequence: AVAVYHGKIS RETGEKLLLA TGLDGSY Antigen sequence identity: Mouse 96%; Rat 93%.
This gene encodes a protein that plays a major role in the bidirectional stimulation of T and B cells. This protein contains an SH2 domain and a short tail. It associates with the signaling lymphocyte-activation molecule, thereby acting as an inhibitor of this transmembrane protein by blocking the recruitment of the SH2-domain-containing signal-transduction molecule SHP-2 to its docking site. This protein can also bind to other related surface molecules that are expressed on activated T, B and NK cells, thereby modifying signal transduction pathways in these cells. Mutations in this gene cause lymphoproliferative syndrome X-linked type 1 or Duncan disease, a rare immunodeficiency characterized by extreme susceptibility to infection with Epstein-Barr virus, with symptoms including severe mononucleosis and malignant lymphoma. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
SH2D1A | |
Polyclonal | |
Unconjugated | |
Sh2d1a | |
DSHP; Duncan disease homolog; Duncan disease SH2-protein; Duncan's disease; EBVS; FLJ18687; FLJ92177; Gm686; HGNC:10820; IMD5; LYP; MTCP1; RGD1562408; RP5-1052M9.3; SAP; SAP/SH2D1A; SH2 domain containing 1A; SH2 domain protein 1A; SH2 domain-containing protein 1A; SH2D1A; signaling lymphocyte activation molecule-associated protein; signaling lymphocytic activation molecule-associated protein; SLAM associated protein/SH2 domain protein 1A; SLAM-associated protein; SLAM-associated protein isoform SAP-2; T cell signal transduction molecule SAP; T-cell signal transduction molecule SAP; Xlp; XLPD; XLPD1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
4068 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60880 | |
Sh2d1a | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 3-29. Recombinant protein control fragment (Product #RP-110143). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction