Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH2D1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586813 Supplier Invitrogen™ Supplier No. PA586813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

This gene encodes a protein that plays a major role in the bidirectional stimulation of T and B cells. This protein contains an SH2 domain and a short tail. It associates with the signaling lymphocyte-activation molecule, thereby acting as an inhibitor of this transmembrane protein by blocking the recruitment of the SH2-domain-containing signal-transduction molecule SHP-2 to its docking site. This protein can also bind to other related surface molecules that are expressed on activated T, B and NK cells, thereby modifying signal transduction pathways in these cells. Mutations in this gene cause lymphoproliferative syndrome X-linked type 1 or Duncan disease, a rare immunodeficiency characterized by extreme susceptibility to infection with Epstein-Barr virus, with symptoms including severe mononucleosis and malignant lymphoma. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D1A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Sh2d1a
Gene Accession No. B2RZ59, O60880, O88890
Gene Alias DSHP; Duncan disease homolog; Duncan disease SH2-protein; Duncan's disease; EBVS; FLJ18687; FLJ92177; Gm686; HGNC:10820; IMD5; LYP; MTCP1; RGD1562408; RP5-1052M9.3; SAP; SAP/SH2D1A; SH2 domain containing 1A; SH2 domain protein 1A; SH2 domain-containing protein 1A; SH2D1A; signaling lymphocyte activation molecule-associated protein; signaling lymphocytic activation molecule-associated protein; SLAM associated protein/SH2 domain protein 1A; SLAM-associated protein; SLAM-associated protein isoform SAP-2; T cell signal transduction molecule SAP; T-cell signal transduction molecule SAP; Xlp; XLPD; XLPD1
Gene Symbols Sh2d1a
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in Human SH2D1A.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20400, 4068, 501502
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.