Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH2D1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563020 Supplier Invitrogen™ Supplier No. PA563020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPYYHGRLTK QDCETLLLKE GVDGNFLL Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 75%.

SH2D1B plays a role in controlling signal transduction through at least four receptors, CD84, CD150, CD229 and CD244, expressed on the surface of professional antigen-presenting cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sh2d1b
Gene Accession No. O14796
Gene Alias Eat2; EAT-2; Eat2a; EAT-2A; EWS/FLI1 activated transcript 2; EWS/FLI1-activated transcript 2; RGD1563935; SH2 domain containing 1B; SH2 domain containing 1B1; SH2 domain protein 1B1; SH2 domain-containing molecule EAT2; SH2 domain-containing protein 1B; Sh2d1b; Sh2d1b1
Gene Symbols Sh2d1b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH2D1B. Recombinant protein control fragment (Product #RP-101476).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117157
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.