Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH2D2A Antibody (OTI4F3), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274153PE
Description
SH2D2A Monoclonal specifically detects SH2D2A in Human samples. It is validated for Flow Cytometry.Specifications
SH2D2A | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
SH2D2A | |
Full length recombinant protein of human SH2D2A (NP_003966) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI4F3 | |
Flow Cytometry | |
F2771, SCAP, SH2 domain containing 2A, SH2 domain protein 2A, SH2 domain-containing adapter protein, T cell specific adapter protein TSAd, T cell-specific adapter protein, T lymphocyte specific adaptor protein, TSAd, TSADSH2 domain-containing protein 2A, VEGF receptor-associated protein, VRAP | |
Mouse | |
Affinity Purified | |
Angiogenesis | |
9047.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction