Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH2D2A Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504381S


Description
SH2D2A Monoclonal antibody specifically detects SH2D2A in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
SH2D2A | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P05154 | |
Mouse | |
Affinity chromatography | |
RUO | |
5104 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI1B8 | |
Unconjugated | |
SERPINA5 | |
F2771; Lad; Lck-associated adapter protein; p56Lck-associated adapter protein; Ribp; Rlk/Itk-binding protein; SCAP; SH2 domain containing 2A; SH2 domain protein 2A; SH2 domain-containing adapter protein; SH2 domain-containing protein 2A; SH22A; SH2D2A; T cell specific adapter protein TSAd; T cell-specific adapter protein; T lymphocyte specific adaptor protein; Tsad; VEGF receptor-associated protein; VRAP | |
Full length human recombinant protein of human SERPINA5 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction