Learn More
SH2D2A Mouse anti-Human, Mouse, Clone: OTI3C7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504372

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Lad (TSAD, TSAd, F2771, SH2D2A, SH2 domain protein 2A) is a 52 kDa cytoplasmic adapter protein that undergoes tyrosine-phosphorylation and influences T cell activation. Lad (SH2D2A) mRNA is present in peripheral blood leukocytes, thymus and spleen, and accumulates in the cytoplasm during T cell activation. The Lad gene maps to chromosome 1q21 in a region where alterations are characteristic to lymphomas.Specifications
SH2D2A | |
Monoclonal | |
Unconjugated | |
SH2D2A | |
F2771, SCAP, TSAD, VRAP | |
Mouse | |
Affinity Chromatography | |
RUO | |
27371, 9047 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3C7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NP31, Q9QXK9 | |
Sh2d2a | |
Full length human recombit protein of human SH2D2A produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.