Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH2D2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537459
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537459 Supplier Invitrogen™ Supplier No. PA537459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 52 kDa. Purity is >95% by SDS-PAGE.

Lad (TSAD, TSAd, F2771, SH2D2A, SH2 domain protein 2A) is a 52 kDa cytoplasmic adapter protein that undergoes tyrosine-phosphorylation and influences T cell activation. Lad (SH2D2A) mRNA is present in peripheral blood leukocytes, thymus and spleen, and accumulates in the cytoplasm during T cell activation. The Lad gene maps to chromosome 1q21 in a region where alterations are characteristic to lymphomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Sh2d2a
Gene Accession No. Q9NP31
Gene Alias F2771; Lad; Lck-associated adapter protein; p56Lck-associated adapter protein; Ribp; Rlk/Itk-binding protein; SCAP; SH2 domain containing 2A; SH2 domain protein 2A; SH2 domain-containing adapter protein; SH2 domain-containing protein 2A; SH22A; SH2D2A; T cell specific adapter protein TSAd; T cell-specific adapter protein; T lymphocyte specific adaptor protein; Tsad; VEGF receptor-associated protein; VRAP
Gene Symbols Sh2d2a
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 218-260 of human Lad.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9047
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.