Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH2D3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519094
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519094 Supplier Invitrogen™ Supplier No. PA519094
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

May play a role in JNK activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SH2D3A
Gene Accession No. Q9BRG2
Gene Alias Novel SH2-containing protein 1; NSP1; SH2 domain containing 3A; SH2 domain-containing 3A; SH2 domain-containing protein 3A; SH2D3A; UNQ175/PRO201
Gene Symbols SH2D3A
Host Species Goat
Immunogen Synthetic peptide sequence (APRAERFEKFQR) corresponding to the internal amino acids of SH2D3A (aa 552-564).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10045
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.