Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SH2D3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566682
Description
Immunogen sequence: WRQLRRSHTEA ALAFEQELKP LMRALDEGAG PCDPGEVALP HVAPMVRLLE GEEVAGPLDE SCERLLRTLH GARHMVRDAP KFRKVAAQRL RGFRPN Highest antigen sequence identity to the following orthologs - mouse 33%, rat 32%.
May play a role in JNK activation.
Specifications
SH2D3A | |
Polyclonal | |
Unconjugated | |
SH2D3A | |
Novel SH2-containing protein 1; NSP1; SH2 domain containing 3A; SH2 domain-containing 3A; SH2 domain-containing protein 3A; SH2D3A; UNQ175/PRO201 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10045 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRG2 | |
SH2D3A | |
Recombinant Human SH2D3A. Recombinant protein control fragment (Product #RP-105310). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction