Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH2D3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583282 Supplier Invitrogen™ Supplier No. PA583282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IEPLRARKWS NSQPADLAHM GRSREDPAGM EASTMPISAL PRTSSDPVLL KAPAPLGTVA DSLRASDGQL QAKAPTKPPR TPSFELPDAS ERP Highest antigen sequence indentity to the following orthologs: Rat - 31%, Mouse - 29%.

May play a role in JNK activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH2D3A
Gene Accession No. Q9BRG2
Gene Alias Novel SH2-containing protein 1; NSP1; SH2 domain containing 3A; SH2 domain-containing 3A; SH2 domain-containing protein 3A; SH2D3A; UNQ175/PRO201
Gene Symbols SH2D3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH2D3A. Recombinant protein control fragment (Product #RP-96518).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10045
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.