Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH2D3C Monoclonal Antibody (2E3)
GREENER_CHOICE

Catalog No. PIMA520895
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520895 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520895 Supplier Invitrogen™ Supplier No. MA520895
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MTAVGRRCPA LGSRGAAGEP EAGSDYVKFS KEKYILDSSP EKLHKELEEE LKLSSTDLRS HAWYHGRIPR EVSETLVQRN GDFLIRDSLT SLGDYVLTCR WRNQALHFKI.

Eph receptor-binding protein which may be a positive regulator of TCR signaling. Binding to BCAR1 is required to induce membrane ruffling and promote EGF-dependent cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D3C
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2E3
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SH2D3C
Gene Accession No. Q8N5H7
Gene Alias Cas/HEF1-associated signal transducer; CHAT; Novel SH2-containing protein 3; NSP3; PRO34088; SH2 domain containing 3C; SH2 domain-containing Eph receptor-binding protein 1; SH2 domain-containing protein 3C; Sh2d3c; Shep1; UNQ272/PRO309/PRO34088
Gene Symbols SH2D3C
Host Species Mouse
Immunogen SH2D3C (NP_005480, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10044
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.