Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH2D3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529448 Supplier Invitrogen™ Supplier No. PA529448
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: SK-N-SH. Predicted reactivity: Mouse (96%), Rat (96%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Eph receptor-binding protein which may be a positive regulator of TCR signaling. Binding to BCAR1 is required to induce membrane ruffling and promote EGF-dependent cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D3C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene SH2D3C
Gene Accession No. Q8N5H7, Q9QZS8
Gene Alias Cas/HEF1-associated signal transducer; CHAT; Novel SH2-containing protein 3; NSP3; PRO34088; SH2 domain containing 3C; SH2 domain-containing Eph receptor-binding protein 1; SH2 domain-containing protein 3C; Sh2d3c; Shep1; UNQ272/PRO309/PRO34088
Gene Symbols SH2D3C
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SH2D3C. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10044, 27387
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.