Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH2D3C Polyclonal Antibody
SH2D3C Polyclonal Antibody
Supplier: Thermo Scientific PA583872
Description
SH2D3C Polyclonal Antibody for Western Blot, IHC (P)

Specifications
SH2D3C | |
Polyclonal | |
Unconjugated | |
SH2D3C | |
Cas/HEF1-associated signal transducer; CHAT; Novel SH2-containing protein 3; NSP3; PRO34088; SH2 domain containing 3C; SH2 domain-containing Eph receptor-binding protein 1; SH2 domain-containing protein 3C; Sh2d3c; Shep1; UNQ272/PRO309/PRO34088 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10044 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N5H7 | |
SH2D3C | |
Recombinant protein corresponding to Human SH2D3C. Recombinant protein control fragment (Product #RP-92892). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction