Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH2D4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545814 Supplier Invitrogen™ Supplier No. PA545814
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IWKKVAEKEE LEQGSRPAPT LEEEKIRSLS SSSRNIQQML ADSINRMKAY Sequence homology: Human: 100%.

SH2D4A inhibits estrogen-induced cell proliferation by competing with PLCG for binding to ESR1, blocking the effect of estrogen on PLCG and repressing estrogen-induced proliferation. SH2D4A may play a role in T-cell development and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D4A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SH2D4A
Gene Accession No. Q9H788
Gene Alias 2210402M20Rik; FLJ20967; PPP1R38; protein phosphatase 1 regulatory subunit 38; protein phosphatase 1, regulatory subunit 38; protein SH(2)A; SH2 domain containing 4A; SH2 domain-containing 4A; SH2 domain-containing protein 4A; SH2A; SH2D4A
Gene Symbols SH2D4A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SH2D4A (aa 145-194).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63898
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.