Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3BGR Monoclonal Antibody (OTI4G5), TrueMAB™, OriGene
SDP

Catalog No. 502543828
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3828 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3828 Supplier OriGene Supplier No. CF812047
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SH3BGR Monoclonal antibody specifically detects SH3BGR in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SH3BGR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4G5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene APOL1
Gene Accession No. O14791
Gene Alias 21-GARP; 21-glutamic acid-rich protein; 5430437A18Rik; SH3 domain binding glutamate rich protein; SH3 domain binding glutamate-rich protein; SH3 domain binding glutamic acid-rich protein; SH3 domain-binding glutamic acid-rich protein; SH3BGR; SH3BGR protein; SH3-binding domain and glutamic acid-rich protein; SH3-binding domain glutamic acid-rich protein
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 28-339 of human APOL1 (NP_003652) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8542
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.