Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3BGR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556539 Supplier Invitrogen™ Supplier No. PA556539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPLLLLGETE PLKLERDCRS PVDPWAAASP DLALACLCHC QDLSSGAFPD RGVL Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 34%.

SH3BGR is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BGR
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sh3bgr
Gene Accession No. P55822
Gene Alias 21-GARP; 21-glutamic acid-rich protein; 5430437A18Rik; SH3 domain binding glutamate rich protein; SH3 domain binding glutamate-rich protein; SH3 domain binding glutamic acid-rich protein; SH3 domain-binding glutamic acid-rich protein; SH3BGR; SH3BGR protein; SH3-binding domain and glutamic acid-rich protein; SH3-binding domain glutamic acid-rich protein
Gene Symbols Sh3bgr
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3BGR. Recombinant protein control fragment (Product #RP-102572).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6450
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.