Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3BGRL Monoclonal Antibody (OTI8F7)
GREENER_CHOICE

Catalog No. PIMA527200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527200 Supplier Invitrogen™ Supplier No. MA527200
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SH3BGRL Monoclonal Antibody for Western Blot, IHC (P)

SH3BGRL belongs to the SH3BGR family. Mutations in predicted EVH1-binding domain of SH3BGRL had a modest effect on suppression of v-Rel transformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BGRL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI8F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SH3BGRL
Gene Accession No. O75368
Gene Alias 1190008F14Rik; epididymis secretory protein Li 115; HEL-S-115; RGD1560520; SH3 domain binding glutamate rich protein like; SH3 domain binding glutamate-rich protein like; SH3 domain binding glutamic acid-rich protein like; SH3 domain-binding glutamic acid-rich-like protein; SH3BGR; Sh3bgrl; SH3-binding domain glutamic acid-rich protein like; similar to SH3 domain binding glutamic acid-rich protein like; zgc:64130; zSH3BGRL
Gene Symbols SH3BGRL
Host Species Mouse
Immunogen Full length human recombinant protein of SH3BGRL produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6451
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.