Learn More
SH3BGRL Mouse anti-Human, Clone: OTI2C7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809728

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SH3BGRL belongs to the SH3BGR family. Mutations in predicted EVH1-binding domain of SH3BGRL had a modest effect on suppression of v-Rel transformation.Specifications
SH3BGRL | |
Monoclonal | |
Unconjugated | |
SH3BGRL | |
HEL-S-115, SH3BGR | |
Mouse | |
Affinity Chromatography | |
RUO | |
6451 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2C7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O75368 | |
SH3BGRL | |
Full length human recombit protein of human SH3BGRL produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.