Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3BGRL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562356 Supplier Invitrogen™ Supplier No. PA562356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FNESQYRGDY DAFFEARENN AVYAFLGLTA PPGSKEAEVQ AKQQA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 98%.

SH3BGRL belongs to the SH3BGR family. Mutations in predicted EVH1-binding domain of SH3BGRL had a modest effect on suppression of v-Rel transformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BGRL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3BGRL
Gene Accession No. O75368
Gene Alias 1190008F14Rik; epididymis secretory protein Li 115; HEL-S-115; RGD1560520; SH3 domain binding glutamate rich protein like; SH3 domain binding glutamate-rich protein like; SH3 domain binding glutamic acid-rich protein like; SH3 domain-binding glutamic acid-rich-like protein; SH3BGR; Sh3bgrl; SH3-binding domain glutamic acid-rich protein like; similar to SH3 domain binding glutamic acid-rich protein like; zgc:64130; zSH3BGRL
Gene Symbols SH3BGRL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3BGRL. Recombinant protein control fragment (Product #RP-101271).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6451
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.