Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3BGRL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561530 Supplier Invitrogen™ Supplier No. PA561530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FEEMDITMSE EQRQWMYKNV PPEKKPTQ Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

SH3BGRL2 gene ontology (GO) annotation includes SH3 domain binding and nuclear membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BGRL2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3BGRL2
Gene Accession No. Q9UJC5
Gene Alias A930014C21Rik; AA408038; B930089I21; FASH3; Fovea-associated SH3 domain-binding protein; SH3 domain binding glutamate rich protein like 2; SH3 domain binding glutamate-rich protein like 2; SH3 domain binding glutamic acid-rich protein like 2; SH3 domain-binding glutamic acid-rich-like protein 2; SH3BGRL2
Gene Symbols SH3BGRL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3BGRL2. Recombinant protein control fragment (Product #RP-100883).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83699
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.