Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3BGRL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104138 Supplier Invitrogen™ Supplier No. PA5104138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SH3BGRL2.

SH3BGRL2 gene ontology (GO) annotation includes SH3 domain binding and nuclear membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BGRL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SH3BGRL2
Gene Accession No. Q8BG73, Q9UJC5
Gene Alias A930014C21Rik; AA408038; B930089I21; FASH3; Fovea-associated SH3 domain-binding protein; SH3 domain binding glutamate rich protein like 2; SH3 domain binding glutamate-rich protein like 2; SH3 domain binding glutamic acid-rich protein like 2; SH3 domain-binding glutamic acid-rich-like protein 2; SH3BGRL2
Gene Symbols SH3BGRL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SH3BGRL2(Accession Q9UJC5), corresponding to amino acid residues M49-P99.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212531, 501026, 83699
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.