Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3BGRL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598958
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598958 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598958 Supplier Invitrogen™ Supplier No. PA598958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SH3BGRL3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

SH3BGRL3 could act as a modulator of glutaredoxin biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BGRL3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SH3BGRL3
Gene Accession No. Q9H299
Gene Alias 1110004L05Rik; AU022188; epididymis secretory protein Li 297; HEL-S-297; P1725; SH3 domain binding glutamate rich protein like 3; SH3 domain binding glutamate-rich protein like 3; SH3 domain binding glutamic acid-rich protein like 3; SH3 domain binding glutamic acid-rich protein-like 3; SH3 domain-binding glutamic acid-rich-like protein 3; SH3 domain-binding protein 1; SH3BGRL3; SH3BGRL3-like protein; SH3BP-1; TIP-B1; TNF inhibitory protein
Gene Symbols SH3BGRL3
Host Species Rabbit
Immunogen Recombinant Human SH3 domain-binding glutamic acid-rich-like protein 3 protein (2-93AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83442
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.