Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3BP1 Monoclonal Antibody (OTI2F4)
GREENER_CHOICE

Catalog No. PIMA527469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527469 Supplier Invitrogen™ Supplier No. MA527469
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SH3BP1 Monoclonal Antibody for Western Blot, IHC (P)

Binds differentially to the SH3 domains of certain proteins of signal transduction pathways. This protein binds preferentially to the ABL1 proto-oncogene, SRC and GRB2. Shows GAP activity for Rac-related proteins but not for Rho- or Ras-related proteins. It inhibits PDGF-induced membrane ruffling mediated by Rac.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3BP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SH3BP1
Gene Accession No. Q9Y3L3
Gene Alias 3bp1; 3BP-1; ARHGAP43; OTTHUMP00000028981; SH3 domain binding protein 1; SH3 domain-binding protein 1; SH3BP1; SH3-domain binding protein 1
Gene Symbols SH3BP1
Host Species Mouse
Immunogen Full length human recombinant protein of SH3BP1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23616
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.